Anti-NF1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: NF1 (neurofibromin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUNB300-155
- NOVUNB300-155SS
- Antigen symbol:
- NF1
- NF1
- Antigen name:
- Neurofibromin 1
- Neurofibromin 1
- Antigen synonyms:
- Neurofibromin|NF1|VRNF|NFNS|WSS|neurofibromin|DKFZp686J1293
- Neurofibromin|NF1|VRNF|NFNS|WSS|neurofibromin|DKFZp686J1293
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4763
- 4763
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together