Anti-Neurokinin B Receptor Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: Neurokinin B Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB300-102
- NOVUNB300-102SS
- Antigen symbol:
- NKBR
- NKBR
- Antigen name:
- Neurokinin B Receptor
- Neurokinin B Receptor
- Antigen synonyms:
- TAC3RL|NKR|NK3R|NK-3R|HH11
- TAC3RL|NKR|NK3R|NK-3R|HH11
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6870
- 6870
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together