Anti-HIST4H4 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB21-2077SS
- NOVUNB21-2077
- Antigen symbol:
- H4
- H4
- Antigen name:
- Histone H4
- Histone H4
- Antigen synonyms:
- H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|HIST1H4B|H4/G|HIST1H4E|H4F2|HIST1H4L|H4/D|HIST4H4|H4FC|H4/E|H4/B|H4FE|HIST1H4A|HIST1H4J|H4FH|H4FM|H4FB|H4/a|HIST1H4K|H4/N|H4/J|HIST2H4|H4/C|HIST1H4F|HIST1H4D|H4FG|HIST1H4I|H4/A|H4FK|H4/H|HIST1H4C|H4FN|H4FD|HIST2H4A
- H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|HIST1H4B|H4/G|HIST1H4E|H4F2|HIST1H4L|H4/D|HIST4H4|H4FC|H4/E|H4/B|H4FE|HIST1H4A|HIST1H4J|H4FH|H4FM|H4FB|H4/a|HIST1H4K|H4/N|H4/J|HIST2H4|H4/C|HIST1H4F|HIST1H4D|H4FG|HIST1H4I|H4/A|H4FK|H4/H|HIST1H4C|H4FN|H4FD|HIST2H4A
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 121504
- 121504
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together