Order Entry
Austria
ContactUsLinkComponent
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-93479SS
  • NOVUNB110-93479
  • Antigen symbol:
  • PDC E1A
  • PDC E1A
  • Antigen name:
  • Pyruvate Dehydrogenase E1-alpha Subunit
  • Pyruvate Dehydrogenase E1-alpha Subunit
  • Antigen synonyms:
  • PDHCE1A|PDHAD|PHE1A|PDHA
  • PDHCE1A|PDHAD|PHE1A|PDHA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 5160
  • 5160
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together