Order Entry
Austria
ContactUsLinkComponent
 
Anti-BAI1 Rabbit Polyclonal Antibody
 
undefined
Anti-BAI1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB110-81586
  • NOVUNB110-81586SS
  • Antigen symbol:
  • ADGRB1
  • ADGRB1
  • Antigen name:
  • Adhesion G Protein-Coupled Receptor B1
  • Adhesion G Protein-Coupled Receptor B1
  • Antigen synonyms:
  • GDAIF|BAI1|R75078|KIAA4089|Brain-Specific Angiogenesis Inhibitor 1
  • GDAIF|BAI1|R75078|KIAA4089|Brain-Specific Angiogenesis Inhibitor 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 575
  • 575
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together