Anti-DEFA5 Mouse Monoclonal Antibody (DyLight® 350) [clone: 8C8]
Catalog # NOVUNB110-60002UV
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:defensin, alpha 5, Paneth cell-specific
- Antigen symbol:DEFA5
- Clonality:Monoclonal
- Clone:8C8
- Conjugation:DyLight® 350
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1670
- Antigen synonyms:HD-5|DEF5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: DEFA5 (defensin, alpha 5, Paneth cell-specific)
Clonality: monoclonal
Clone: 8C8
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human