Anti-CD36 Mouse Monoclonal Antibody (DyLight® 488) [clone: D-2712]
NOVUNB110-59724G
- Antibody type:Primary
- Antigen name:CD36 molecule
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:D-2712
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgA kappa
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:948
- Antigen synonyms:GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone: D-2712
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat