Order Entry
Austria
ContactUsLinkComponent
Anti-SAT1 Rabbit Polyclonal Antibody
Anti-SAT1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
Anti-SAT1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-41622
  • NOVUNB110-41622SS
  • Antigen symbol:
  • SAT1
  • SAT1
  • Antigen name:
  • Spermidine/spermine N1-acetyltransferase 1
  • Spermidine/spermine N1-acetyltransferase 1
  • Antigen synonyms:
  • SSAT-1|SSAT|Spermidine/spermine N(1)-acetyltransferase 1|Polyamine N-acetylt|Puterscine acetyltransferase|EC 2.3.1.57|Diamine acetytransferase 1
  • SSAT-1|SSAT|Spermidine/spermine N(1)-acetyltransferase 1|Polyamine N-acetylt|Puterscine acetyltransferase|EC 2.3.1.57|Diamine acetytransferase 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6303
  • 6303
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together