Anti-5-HT3A Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56351
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine receptor 3a
- Antigen symbol:5HT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3359
- Antigen synonyms:5-HT3A|5HT3 receptor|5-HT3|5 HT3|5HT3|79246|Htr3|5 HT3A|Htr3a|5HT3A|5 HT3 receptor|5HT
- Storage buffer:PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
The 5-HT3A Antibody from Novus Biologicals is a rabbit polyclonal antibody to 5-HT3A. This antibody reacts with human, mouse, rat. The 5-HT3A Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Simple Western,
Type: Primary
Antigen: 5-HT3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat