Anti-CDK5R1 Rabbit Polyclonal Antibody
NOVUNB100-56153
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 5 regulatory subunit 1
- Antigen symbol:CDK5R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:8851
- Antigen synonyms:p35nck5a|CDK5R|p35|p23|CDK5P35|NCK5A|p25
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
Type: Primary
Antigen: CDK5R1 (cyclin-dependent kinase 5, regulatory subunit 1 (p35))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan