Anti-CD40 Rabbit Polyclonal Antibody
NOVUNB100-56127
- Antibody type:Primary
- Antigen name:CD40 molecule, TNF receptor superfamily member 5
- Antigen symbol:CD40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:958
- Antigen synonyms:p50|Bp50|TNFRSF5|CDW40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: