Anti-BCL2L1 Rabbit Polyclonal Antibody
NOVUNB100-56104
- Antibody type:Primary
- Antigen name:BCL2-like 1
- Antigen symbol:BCL2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:598
- Antigen synonyms:BCLXL|BCLX|BCLXS|BCL2L|BCL-XL/S|Bcl-X|PPP1R52|bcl-xS|bcl-xL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat