Anti-SLC31A1/CTR1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-402F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 31 (copper transporter), member 1
- Antigen symbol:Solute carrier family 31, member 1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1317
- Antigen synonyms:COPT1|CTR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC31A1/CTR1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: