Anti-RAD21 Mouse Monoclonal Antibody [clone: CM110-2C10]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: monoclonal
Clone: CM110-2C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNB100-386
- NOVUNB100-386SS
- Antigen symbol:
- RAD21
- RAD21
- Antigen name:
- RAD21 homolog
- RAD21 homolog
- Antigen synonyms:
- SCC1|hHR21|KIAA0078|FLJ40596|RAD21|MCD1|NXP1|FLJ25655|HR21|NXP-1|HRAD21
- SCC1|hHR21|KIAA0078|FLJ40596|RAD21|MCD1|NXP1|FLJ25655|HR21|NXP-1|HRAD21
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- CM110-2C10
- CM110-2C10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 5885
- 5885
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together