Order Entry
Austria
ContactUsLinkComponent
Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]
Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-353
  • NOVUNB100-353SS
  • Antigen symbol:
  • BubR1
  • BubR1
  • Antigen name:
  • Mitotic checkpoint serine/threonine-protein kinase BUB1
  • Mitotic checkpoint serine/threonine-protein kinase BUB1
  • Antigen synonyms:
  • mitotic checkpoint kinase MAD3L|MAD3L|MAD3/BUB1-related protein kinase|hBUBR1|SSK1|mitotic checkpoint serine/threonine-protein kinase BUB1 beta|BUBR1|BUB1beta|Bub1A
  • mitotic checkpoint kinase MAD3L|MAD3L|MAD3/BUB1-related protein kinase|hBUBR1|SSK1|mitotic checkpoint serine/threonine-protein kinase BUB1 beta|BUBR1|BUB1beta|Bub1A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8G1
  • 8G1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 701
  • 701
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together