Anti-TERT Mouse Monoclonal Antibody [clone: 2D8]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: monoclonal
Clone: 2D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-297
- NOVUNB100-297SS
- Antigen symbol:
- TERT
- TERT
- Antigen name:
- Telomerase Reverse Transcriptase
- Telomerase Reverse Transcriptase
- Antigen synonyms:
- TRT|telomerase-associated protein 2|hEST2|EST2|DKCA2|DKCB4|TP2|TCS1|telomerase catalytic subunit|hTRT
- TRT|telomerase-associated protein 2|hEST2|EST2|DKCA2|DKCB4|TP2|TCS1|telomerase catalytic subunit|hTRT
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2D8
- 2D8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7015
- 7015
- Isotype:
- IgM
- IgM
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together