Order Entry
Austria
ContactUsLinkComponent
Anti-Tuberin Rabbit Polyclonal Antibody
Anti-Tuberin Rabbit Polyclonal Antibody
Catalog # BSBTPA2175
Supplier:  Boster Bio
undefined
Anti-Tuberin Rabbit Polyclonal Antibody
Catalog # BSBTPA2175
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tuberous Sclerosis Complex 2
  • Antigen symbol:
    TSC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    TSC4|TSC2|Tuberous sclerosis 2 protein|Tuberin
  • Amino acid number:
    1605 to 1620
  • UniProtKB:
    P49815
  • Sequence:
    QFTYCWHDDIMQAVFH
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Tuberin (1605-1620aa QFTYCWHDDIMQAVFH), identical to the related mouse and rat sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Tuberin(TSC2) detection. Tested with WB in Human;Mouse;Rat.

Tuberous sclerosis 2 protein, also known as TSC2 or Tuberin is a protein that is in humans. The chromosomal location of this gene is 16p13.3. Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. This gene involved in microtubule-mediated protein transport, but this seems to be due to unregulated mTOR signaling. It stimulates weakly the intrinsic GTPase activity of the Ras-related proteins RAP1A and RAB5 in vitro.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Tuberin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat