Order Entry
Austria
ContactUsLinkComponent
Anti-ABCA4 Rabbit Polyclonal Antibody
Anti-ABCA4 Rabbit Polyclonal Antibody
Catalog # BSBTPA2065
Supplier:  Boster Bio
undefined
Anti-ABCA4 Rabbit Polyclonal Antibody
Catalog # BSBTPA2065
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP Binding Cassette, sub-family A, member 4
  • Antigen symbol:
    ABCA4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    ABCA4
  • Amino acid number:
    1893 - 1906
  • Sequence:
    TLLVQRHFFLSQWI
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human ABCA4 (1893-1906aa TLLVQRHFFLSQWI)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Retinal-specific ATP-binding cassette transporter(ABCA4) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ABCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human