Order Entry
Austria
ContactUsLinkComponent
 
Anti-LPP Rabbit Polyclonal Antibody
  BSBTPA2040
undefined
Anti-LPP Rabbit Polyclonal Antibody
  BSBTPA2040

 

  • Antibody type:
    Primary
  • Antigen name:
    LIM Domain Containing Preferred Translocation Partner In Lipoma
  • Antigen symbol:
    LPP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    291 - 307
  • Sequence:
    PNQGRYYEGYYAAGPGY
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human LPP (291-307aa PNQGRYYEGYYAAGPGY).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Lipoma-preferred partner (LPP) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: LPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human