Order Entry
Austria
ContactUsLinkComponent
Anti-PERK Rabbit Polyclonal Antibody
Anti-PERK Rabbit Polyclonal Antibody
  BSBTPA2026
undefined
Anti-PERK Rabbit Polyclonal Antibody
  BSBTPA2026

 

  • Antibody type:
    Primary
  • Antigen name:
    PRKR-like endoplasmic reticulum kinase
  • Antigen symbol:
    PERK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    PRKR-like endoplasmic reticulum kinase|HsPEK|Pancreatic eIF2-alpha kinase|Eukaryotic translation initiation factor 2-alpha kinase 3
  • Amino acid number:
    163 - 176
  • Sequence:
    QWDQDRESMETVPF
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human PERK (163-176aa QWDQDRESMETVPF).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Eukaryotic translation initiation factor 2-alpha kinase 3 (EIF2AK3) detection. Tested with WB, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PERK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human