Anti-Cytochrome P450 Reductase Rabbit Polyclonal Antibody
BSBTPA1952
- Antibody type:Primary
- Antigen name:P450 (cytochrome) oxidoreductase
- Antigen symbol:POR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:CPR|DKFZp686G04235|P450R|CYPOR|FLJ26468
- Amino acid number:659 - 677
- Sequence:DYIKKLMTKGRYSLDVWS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Cytochrome P450 Reductase (659-677aa DYIKKLMTKGRYSLDVWS), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for NADPH--cytochrome P450 reductase (POR) detection. Tested with WB, IHC-P, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Cytochrome P450 Reductase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat