Order Entry
Austria
ContactUsLinkComponent
Anti-PTCH2 Rabbit Polyclonal Antibody
Anti-PTCH2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1859
Supplier:  Boster Bio
undefined
Anti-PTCH2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1859
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Patched 2
  • Antigen symbol:
    PTCH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    PTC2
  • Amino acid number:
    885 - 901
  • Sequence:
    WLHDKYDTTGENLRIPP
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human PTCH2 (885-901aa WLHDKYDTTGENLRIPP), different from the related mouse sequence by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Protein patched homolog 2 (PTCH2) detection. Tested with WB in Human; Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse