Anti-Coxsackie Adenovirus Receptor Rabbit Polyclonal Antibody
BSBTPA1852
- Antibody type:Primary
- Antigen name:Coxsackie Adenovirus Receptor
- Antigen symbol:CXADR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:313 - 328
- Sequence:YSKTQYNQVPSEDFER
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Coxsackie Adenovirus Receptor (313-328aa YSKTQYNQVPSEDFER), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Coxsackievirus and adenovirus receptor (CXADR) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Coxsackie Adenovirus Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat