Anti-ADAM17 Rabbit Polyclonal Antibody
BSBTPA1844
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 17
- Antigen symbol:ADAM17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:TACE|Snake venom-like protease|ADAM18|CSVP|TNF-alpha-converting enzyme|CD156B|CD156b antigen|TNF-alpha convertase|A disintegrin and metalloproteinase domain 17|NISBD|NISBD1
- Amino acid number:806 - 824
- Sequence:AASFKLQRQNRVDSKETEC
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ADAM17 (806-824aa AASFKLQRQNRVDSKETEC), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Disintegrin and metalloproteinase domain-containing protein 17 (ADAM17) detection. Tested with WB in Human; Mouse; Rat.
- Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ADAM17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat