Anti-Cdk7/CAK Mouse Monoclonal Antibody [clone: IMD-26]
BSBTMA1022
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 7
- Antigen symbol:CDK7
- Clonality:Monoclonal
- Clone:IMD-26
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CAK1|MO15|Cell division protein kinase 7|CDK7|CAK|TFIIH basal transcription factor complex kinase subunit|39 kDa protein kinase|P39 Mo15|STK1|CDK-activating kinase
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human Cdk7 protein.
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Cdk7/CAK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human