Order Entry
Austria
ContactUsLinkComponent
Anti-CDK6 Mouse Monoclonal Antibody [clone: IML-6]
Anti-CDK6 Mouse Monoclonal Antibody [clone: IML-6]
Catalog # BSBTMA1021
Supplier:  Boster Bio
Anti-CDK6 Mouse Monoclonal Antibody [clone: IML-6]
Catalog # BSBTMA1021
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase 6
  • Antigen symbol:
    CDK6
  • Clonality:
    Monoclonal
  • Clone:
    IML-6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    Serine/threonine-protein kinase PLSTIRE|Cyclin-dependent kinase 6|MCPH12|PLSTIRE|Cell division protein kinase 6|CDKN6|CDK6
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human Cdk6 protein.
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: CDK6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat