Order Entry
Austria
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (Biotin)
Anti-IgG Goat Polyclonal Antibody (Biotin)
Supplier:  Biotium
Certificates
Anti-IgG Goat Polyclonal Antibody (Biotin)
Supplier:  Biotium

Specifications

  • Catalog No:
  • 20188.
  • BTIU20188-1
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Rabbit IgG
  • Rabbit IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Antibody fragment:
  • F(ab')2
  • F(ab')2
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together