Order Entry
Austria
ContactUsLinkComponent
Anti-SLC10A2/ASBT Goat Polyclonal Antibody
Anti-SLC10A2/ASBT Goat Polyclonal Antibody
Catalog # ABCAAB134837-50
Supplier:  Abcam
undefined
Anti-SLC10A2/ASBT Goat Polyclonal Antibody
Catalog # ABCAAB134837-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 10 (sodium/bile acid cotransporter), member 2
  • Antigen symbol:
    Solute carrier family 10, member 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Goat polyclonal to SLC10A2/ASBT.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SLC10A2/ASBT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human