Order Entry
Austria
ContactUsLinkComponent
Anti-GPCR GPR43 Rabbit Polyclonal Antibody
Anti-GPCR GPR43 Rabbit Polyclonal Antibody
  ABCAAB124272-50
 :  Abcam
undefined
Anti-GPCR GPR43 Rabbit Polyclonal Antibody
  ABCAAB124272-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Free fatty acid receptor 2
  • Antigen symbol:
    FFAR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Rabbit polyclonal to GPCR GPR43.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GPCR GPR43
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;African green monkey