Order Entry
Austria
ContactUsLinkComponent
Anti-ELOVL7 Rabbit Polyclonal Antibody
Anti-ELOVL7 Rabbit Polyclonal Antibody
  ABCAAB106633-100
 :  Abcam
undefined
Anti-ELOVL7 Rabbit Polyclonal Antibody
  ABCAAB106633-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    ELOVL fatty acid elongase 7
  • Antigen symbol:
    ELOVL7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal to ELOVL7.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ELOVL7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human