Order Entry
Austria
ContactUsLinkComponent
Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 1A5]
Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 1A5]
Catalog # ABCAAB97426-50
Supplier:  Abcam
undefined
Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 1A5]
Catalog # ABCAAB97426-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialic acid binding Ig-like lectin 5
  • Antigen symbol:
    SIGLEC5
  • Clonality:
    Monoclonal
  • Clone:
    1A5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [1A5] to SIGLEC5.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 1A5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human