Order Entry
Austria
ContactUsLinkComponent
Anti-Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 Rabbit Polyclonal Antibody
Anti-Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 Rabbit Polyclonal Antibody
Catalog # ABCAAB65266-200
Supplier:  Abcam
undefined
Anti-Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 Rabbit Polyclonal Antibody
Catalog # ABCAAB65266-200
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium channel L type DHPR alpha 2 subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    200 µl

Specifications

About this item

Rabbit polyclonal to Calcium channel L type DHPR alpha 2 subunit/CACNA2D1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Calcium channel L type DHPR alpha 2 subunit/CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human