Anti-CCNE1 Rabbit polyclonal antibody unconjugated
Catalog # BIRBORB99278-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Modification:Phospho-Thr395
- Storage buffer:phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:48 kDa
- Storage temperature:Cool
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthesized peptide derived from human Cyclin E1 around the phosphorylation site of Threonine 395.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human