Anti-IgA/IgG1/IgG2a/IgG2b/IgG3/IgM Goat Polyclonal Antibody
BIRBORB98163-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgA/IgG1/IgG2a/IgG2b/IgG3/IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Mouse IgG1 IgG2a IgG2b IgG3 IgA IgM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse