Anti-HP Mouse Monoclonal Antibody
BIRBORB95643-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Haptoglobin
- Antigen symbol:HP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Storage buffer:HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µg
- Pk:50 µG
Immunochemistry: 1:200; Western Blot: 1:500
Type: Primary
Antigen: HP (haptoglobin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human