Anti-SERPINH1 Rabbit Polyclonal Antibody
Catalog # BIRBORB88006-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade H, member 1
- Antigen symbol:SERPINH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage buffer:PBS, 30% glycerol, 0,5% BSA, 0,01% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10-20 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunoprecipitation: 10-20 ?g/ml; Western Blot: 0.5-4 ?g/ml
Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: