Anti-TCP1 Beta Mouse Monoclonal Antibody
Catalog # BIRBORB87514-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:TCP1 beta
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Phosphate-buffered solution, pH 7,2, 0,09% sodium azide at 0,5 mg/ml.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 15 ?g/ml
Type: Primary
Antigen: TCP1 Beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat