Anti-Dopamine D2 Receptor Rabbit Polyclonal Antibody
BIRBORB85442-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Dopamine receptor D2
- Antigen symbol:DRD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human DRD2
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 3-4 ?g/ml
Type: Primary
Antigen: Dopamine D2 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human