Anti-F2RL3 Rabbit Polyclonal Antibody
Catalog # BIRBORB84952-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:coagulation factor II (thrombin) receptor-like 3
- Antigen symbol:F2RL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 16 amino acid peptide from N-terminal extracellular domain of human F2RL3/PAR4
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 6-13 ?g/ml
Type: Primary
Antigen: F2RL3 (coagulation factor II (thrombin) receptor-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human