Anti-Esherichia coli heat-labile enterotoxin B-chain Mouse Monoclonal Antibody
Catalog # BIRBORB79402-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Esherichia coli heat-labile enterotoxin B-chain
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Esherichia coli heat-labile enterotoxin B-chain
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human