Anti-CCNA2 Rabbit polyclonal antibody unconjugated
Catalog # BIRBORB77404-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cyclin A2
- Antigen symbol:CCNA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Peptide
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:1000-1:100000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human