Anti-PP2A C Mouse Monoclonal Antibody [clone: 2A10]
BIRBORB76780-0.1
: Biorbyt
- Antibody type:Primary
- Antigen symbol:PP2A C
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:C-terminal peptide Methyl-Leu309 of human PP2A catalytic
- Purification:purified by Protein G chromatography
- Size:100 μL
- Pk:0,1 mL
Western Blot: 1:500
Type: Primary
Antigen: PP2A C
Clonality: monoclonal
Clone: 2A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human