Anti-Nesprin 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB76724-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Spectrin Repeat Containing Nuclear Envelope Protein 1
- Antigen symbol:SYNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:antiserum
- Storage buffer:PBS, 0,025% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic polypeptide GRSTPNRQKTPRGKC residues 3212-3226conjugated to KLH
- Purification:purified by affinity chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Western Blot: 1:500-1:750; Immunofluorescence: 1:25-1:50
Type: Primary
Antigen: Nesprin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: