Anti-ACTB Rabbit Polyclonal Antibody
Catalog # BIRBORB74081-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Actin beta
- Antigen symbol:ACTB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Concentration:0,2 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human