Anti-RAB3C Rabbit Polyclonal Antibody
BIRBORB69874-100
: Biorbyt
- Antibody type:Primary
- Antigen name:RAB3C, member RAS oncogene family
- Antigen symbol:RAB3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:0,94 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: RAB3C (RAB3C, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human