Anti-AKT2 Mouse Monoclonal Antibody
BIRBORB69223-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Protein Kinase B beta
- Antigen symbol:PKBB
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites fluid
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Purified recombinant fragment of human AKT2 expressed in E. Coli.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10000
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: