Anti-P2RX4 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131558-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:purinergic receptor P2X, ligand-gated ion channel, 4
- Antigen symbol:P2RX4
- Clonality:Polyclonal
- Conjugation:RBITC (Rhodamine B Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:43 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: P2RX4 (purinergic receptor P2X, ligand-gated ion channel, 4)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: