Anti-CXCL10 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131321-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) ligand 10
- Antigen symbol:CXCL10
- Clonality:Polyclonal
- Conjugation:RBITC (Rhodamine B Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:10 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CXCL10 (chemokine (C-X-C motif) ligand 10)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat