Anti-Mast Cell Chymase Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)
Catalog # BIRBORB130026-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Chymase, Mast Cell
- Antigen symbol:CMA
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:27 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human Ceruloplasmin.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Mast Cell Chymase
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat