Order Entry
Austria
ContactUsLinkComponent
Anti-MEF2C Rabbit Polyclonal Antibody
Anti-MEF2C Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-MEF2C Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB129685-100
  • BIRBORB129685-200
  • Antigen symbol:
  • MEF2C
  • MEF2C
  • Antigen name:
  • Myocyte enhancer factor 2C
  • Myocyte enhancer factor 2C
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together